Triethanolamine (trolamine) 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: Tris-(2-hydroxyethyl)amine, 2,2',2''-Nitrilotriethanol

L04486-0E L04486-22 L04486-36
CAAAAL04486-0EEA 146.62 CAD
CAAAAL04486-0E CAAAAL04486-22 CAAAAL04486-36
Triethanolamine (trolamine) 98+%
Triethanolamine (Trolamine)

Protein purification reagent


Purity: 98+% CAS No.: 102-71-6 Molecular Formula: C6H15NO3 Formula Weight: 149.19 Flash Point: 185°(365°F) Refractive Index: 1.4850 Odor: Ammonia-like Melting Point: 19-21° Boiling Point: 206-207°/15mm Beilstein Reference: 1699263 Merck Reference: 14,9665 MDL No.: MFCD00002855

Formula: C₆H₁₅NO₃
MW: 149.19 g/mol
Boiling Pt: 206…207 °C/15 mm
Melting Pt: 19…21 °C
Density: 1.124
Flash Pt: 185 °C(365 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002855
CAS Number: 102-71-6
EINECS: 203-049-8
Merck Index: 14,9665
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c181d3e80375aec7c34e17766247484e1c3d3257&vid=40c3d3d0c0162ac22208cadb8d5254a7cf0b340a&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR