5-Aminouracil 97%

Supplier: Thermo Scientific Chemicals
Warning

L04452-06 L04452-14
CAAAAL04452-06EA 57.51 CAD
CAAAAL04452-06 CAAAAL04452-14
5-Aminouracil 97%
5-Aminouracil

Purity: 97% CAS No.: 932-52-5 Molecular Formula: C4H5N3O2 Formula Weight: 127.10 Melting Point: 300° Beilstein Reference: 127250

Formula: C₄H₅N₃O₂
Boiling Pt: 377 °C (1013 hPa)
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00006025
CAS Number: 932-52-5
EINECS: 213-252-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR