4-Hydroxythiophenol 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Hydroxybenzenethiol, p-Hydroxythiophenol, 1-Hydroxy-4-mercaptobenzene, 4-Mercaptophenol, 4-Hydroxyphenyl mercaptan, 4-Hydroxybenzene-1-thiol

L04429-06 L04429-14
CAAAAL04429-06EA 82.79 CAD
CAAAAL04429-06 CAAAAL04429-14
4-Hydroxythiophenol 97%
4-Hydroxythiophenol

Purity: 97% CAS No.: 637-89-8 Molecular Formula: C6H6OS Formula Weight: 126.18 Flash Point: 110°(230°F) Odor: Unpleasant Boiling Point: 256° Beilstein Reference: 2039306 UN No.: 1759 MDL No.: MFCD00004850

Formula: C₆H₆OS
MW: 126.18 g/mol
Boiling Pt: 256
Melting Pt: 33…37 °C
Density: 1.255
Flash Pt: 110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004850
CAS Number: 637-89-8
EINECS: 211-307-6
UN: 2923
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR