Octyltriethoxysilane 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Triethoxy(octyl)silane, Triethoxyoctylsilane

L04407-0B L04407-18 L04407-30
CAAAAL04407-0BEA 214.88 CAD
CAAAAL04407-0B CAAAAL04407-18 CAAAAL04407-30
Octyltriethoxysilane 95%
Octyltriethoxysilane

Purity: 95% CAS No.: 2943-75-1 Molecular Formula: CH3(CH2)7Si(OCH2CH3)3 Formula Weight: 276.49 Flash Point: 109°(228°F) Refractive Index: 1.4170 Melting Point: -40° Boiling Point: 265° Beilstein Reference: 2325287 MDL No.: MFCD00039883

Formula: C₁₄H₃₂O₃Si
Boiling Pt: 265 °C
Melting Pt: < –40 °C
Density: 0.878
Flash Pt: 109 °C(228 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039883
CAS Number: 2943-75-1
EINECS: 220-941-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR