Isovaleric anhydride 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L04385-06
L04385-14
CAAAAL04385-06EA
38.24
CAD
CAAAAL04385-06
CAAAAL04385-14
Isovaleric anhydride 95%
Isovaleric anhydride
Purity: 95%
CAS No.: 1468-39-9
Molecular Formula: C10H18O3
Formula Weight: 186.25
Flash Point: 91°(195°F)
Refractive Index: 1.4165
Melting Point: -65°
Boiling Point: 103-104°/24mm
Beilstein Reference: 1635850
UN No.: 3265
MDL No.: MFCD00015051
Formula:
C₁₀H₁₈O₃ Boiling Pt: 103…104 °C/24 mm Melting Pt: –65 °C Density: 0.927 Flash Pt: 91 °C (195 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00015051 CAS Number: 1468-39-9 EINECS: 215-994-3 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...