2,6-Dimethoxy-3-nitrobenzoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

L04366-06 L04366-14
CAAAAL04366-06EA 78.84 CAD
CAAAAL04366-06 CAAAAL04366-14
2,6-Dimethoxy-3-nitrobenzoic acid 97%
2,6-Dimethoxy-3-nitrobenzoic acid

Purity: 97% CAS No.: 55776-17-5 Molecular Formula: C9H9NO6 Formula Weight: 227.17 Melting Point: 130-132° Beilstein Reference: 2657939

Formula: C₉H₉NO₆
MW: 227.17 g/mol
Melting Pt: 130…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00017324
CAS Number: 55776-17-5
EINECS: 259-814-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR