3-Hydroxybenzhydrazide 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L04357-03
L04357-06
CAAAAL04357-03EA
41.4
CAD
CAAAAL04357-03
CAAAAL04357-06
3-Hydroxybenzhydrazide 98+%
3-Hydroxybenzhydrazide
Purity: 98+%
CAS No.: 5818-06-4
Molecular Formula: C7H8N2O2
Formula Weight: 152.15
Melting Point: 157-159°
Beilstein Reference: 638933
Formula:
C₇H₈N₂O₂ Melting Pt: 157…159 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014759 CAS Number: 5818-06-4 EINECS: 227-389-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...