Chlorocyclohexane 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Cyclohexyl chloride
L04349-0B
L04349-30
CAAAAL04349-0BEA
202.24
CAD
CAAAAL04349-0B
CAAAAL04349-30
Chlorocyclohexane 98%
Chlorocyclohexane
Purity: 98%
CAS No.: 542-18-7
Molecular Formula: C6H11Cl
Formula Weight: 118.61
Flash Point: 28°(82°F)
Refractive Index: 1.4620
Odor: Unpleasant
Melting Point: -44°
Boiling Point: 141-143°
Beilstein Reference: 1900796
Merck Reference: 14,2732
UN No.: 1993
MDL No.: MFCD00003822
Formula:
C₆H₁₁Cl MW: 118.61 g/mol Boiling Pt: 142 Melting Pt: -44 Density: 1.000 Flash Pt: 28 °C (82 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003822 CAS Number: 542-18-7 EINECS: 208-806-6 UN: 1993 ADR: 3 III Merck Index: 14,2732 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...