1,2,3-Trichloropropane 98+%

Supplier: Thermo Scientific Chemicals
Danger

L04312-0B L04312-30
CAAAAL04312-0BEA 157.84 CAD
CAAAAL04312-0B CAAAAL04312-30
1,2,3-Trichloropropane 98+%
1,2,3-Trichloropropane

Purity: 98+% CAS No.: 96-18-4 Molecular Formula: C3H5Cl3 Formula Weight: 147.43 Flash Point: 74°(165°F) Refractive Index: 1.4822 Melting Point: -14° Boiling Point: 156-158° Beilstein Reference: 1732068 UN No.: 2810 MDL No.: MFCD00000946

Formula: ClH₂CCHClCH₂Cl
Boiling Pt: 156…158 °C
Melting Pt: –14 °C
Density: 1.385
Flash Pt: 74 °C (165 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000946
CAS Number: 96-18-4
EINECS: 202-486-1
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR