(1-Tetradecyl)triphenylphosphonium bromide 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Triphenyl(tetradecyl)phosphonium bromide, Tetradecyltriphenylphosphonium bromide

L04311-14 L04311-22
CAAAAL04311-14EA 136.35 CAD
CAAAAL04311-14 CAAAAL04311-22
(1-Tetradecyl)triphenylphosphonium bromide 97%
(1-Tetradecyl)triphenylphosphonium bromide

Purity: 97% CAS No.: 25791-20-2 Molecular Formula: CH3(CH2)13P(C6H5)3Br Formula Weight: 539.59 Melting Point: 87-89° Beilstein Reference: 3582243

Formula: H₃C(CH₂)₁₃P(C₆H₅)₃Br
MW: 539.58 g/mol
Melting Pt: 87…89 °C
Storage Temperature: Ambient
MDL Number: MFCD00051857
CAS Number: 25791-20-2
EINECS: 247-267-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR