Acrylic acid 99% stabilised
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Prop-2-enoic acid
Stabilization:
∼200 ppm Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
L04280-22
L04280-36
CAAAAL04280-22EA
124.63
CAD
CAAAAL04280-22
CAAAAL04280-36
Acrylic acid 99% stabilised
Acrylic acid
Purity: 99%
CAS No.: 79-10-7
Molecular Formula: C3H4O2
Formula Weight: 72.06
Flash Point: 54°(130°F)
Refractive Index: 1.4202
Odor: Pungent
Melting Point: ca 13°
Boiling Point: 138-139°
Beilstein Reference: 635743
Merck Reference: 14,130
UN No.: 2218
MDL No.: MFCD00004367
Formula:
C₃H₄O₂ MW: 72.06 g/mol Boiling Pt: 138…139 °C Melting Pt: ∼ 13 °C Density: 1.051 Flash Pt: 54 °C (130 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004367 CAS Number: 79-10-7 EINECS: 201-177-9 UN: 2218 ADR: 8 II Merck Index: 14,130 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...