2,6-Diaminotoluene 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,3-Diamino-2-methylbenzene, Toluene-2,6-diamine

L04278-06 L04278-14
CAAAAL04278-06EA 54.98 CAD
CAAAAL04278-06 CAAAAL04278-14
2,6-Diaminotoluene 97%
2,6-Diaminotoluene

Purity: 97% CAS No.: 823-40-5 Molecular Formula: C7H10N2 Formula Weight: 122.17 Melting Point: 104-106° Beilstein Reference: 2079476 UN No.: 2811 MDL No.: MFCD00007800

Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Boiling Pt: 280…282 °C (1013 hPa)
Melting Pt: 104…106 °C
Storage Temperature: Ambient
MDL Number: MFCD00007800
CAS Number: 823-40-5
EINECS: 212-513-9
UN: 3077
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR