Isopropyl palmitate tech. 85%, Technical Grade
Supplier: Thermo Scientific Chemicals
Synonyms:
Hexadecylic acid isopropyl ester, Palmitic acid isopropyl ester, Hexadecanoic acid isopropyl ester, Isopropyl Palmitate, Isopropyl hexadecanoate
L04276-0F
L04276-AK
L04276-AU
CAAAAL04276-0FEA
189.6
CAD
CAAAAL04276-0F
CAAAAL04276-AK
CAAAAL04276-AU
Isopropyl palmitate tech. 85%, Technical Grade
Isopropyl palmitate
Purity: 85%
CAS No.: 142-91-6
Molecular Formula: CH3(CH2)14CO2CH(CH3)2
Formula Weight: 298.51
Flash Point: 183°(361°F)
Refractive Index: 1.4380
Melting Point: 11-13°
Boiling Point: 160°/2mm
Beilstein Reference: 1786567
MDL No.: MFCD00008993
Formula:
C₁₉H₃₈O₂ MW: 298.51 g/mol Boiling Pt: 160 Melting Pt: 13.5 Density: 0.852 Flash Pt: 183 °C (361 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008993 CAS Number: 142-91-6 EINECS: 205-571-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...