1,12-Diaminododecane 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N12N, Dodecamethylendiamine, 1,12-Dodecanediamine

L04258-06 L04258-14
CAAAAL04258-06EA 59.41 CAD
CAAAAL04258-06 CAAAAL04258-14
1,12-Diaminododecane 98+%
1,12-Diaminododecane

Purity: 98+% CAS No.: 2783-17-7 Molecular Formula: C12H28N2 Formula Weight: 200.37 Flash Point: 155°(311°F) Melting Point: 68-72° Beilstein Reference: 1742765 UN No.: 3259 MDL No.: MFCD00008155

Formula: C₁₂H₂₈N₂
MW: 200.37 g/mol
Boiling Pt: 275 °C (1013 hPa)
Melting Pt: 68…72 °C
Flash Pt: 155 °C (311 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008155
CAS Number: 2783-17-7
EINECS: 220-489-6
UN: 3259
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR