Oil of castor

Supplier: Thermo Scientific Chemicals

Synonyms: Castor oil, Ricinus oil, Oleum Ricini s. Castoris, Oil of Palma Christi

L04224-0F L04224-AK L04224-AU
CAAAAL04224-0FEA 227.52 CAD
CAAAAL04224-0F CAAAAL04224-AK CAAAAL04224-AU
Oil of castor
Oil of castor

CAS No.: 8001-79-4 Flash Point: 230°(446°F) Refractive Index: 1.4780 Boiling Point: 313° Merck Reference: 14,1898 Optical Rotation: +5° (c=5 in ethanol)

Boiling Pt: 313 °C
Melting Pt: –10 °C
Density: 0.961
Flash Pt: 230 °C(446 °F)
Storage Temperature: Ambient
MDL Number: MFCD00130746
CAS Number: 8001-79-4
EINECS: 232-293-8
Merck Index: 14,1898

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR