Methyldiphenylsilane 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Diphenylmethylsilane
L04211-06
L04211-14
CAAAAL04211-06EA
64.46
CAD
CAAAAL04211-06
CAAAAL04211-14
Methyldiphenylsilane 97%
Methyldiphenylsilane
Purity: 97%
CAS No.: 776-76-1
Molecular Formula: C13H14Si
Formula Weight: 198.34
Flash Point: 120°(248°F)
Refractive Index: 1.5720
Odor: Odorless
Boiling Point: 266-267°
Beilstein Reference: 1867183
MDL No.: MFCD00008255
Formula:
C₁₃H₁₄Si MW: 198.34 g/mol Boiling Pt: 266…267 °C Density: 1,001 Flash Pt: 120 °C (248 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008255 CAS Number: 776-76-1 EINECS: 212-281-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...