Propionic acid 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Propanoic acid
L04210-0B
L04210-0E
L04210-22
CAAAAL04210-0BEA
62.25
CAD
CAAAAL04210-0B
CAAAAL04210-0E
CAAAAL04210-22
Propionic acid 99%
Propionic acid
Purity: 99%
CAS No.: 79-09-4
Molecular Formula: C3H6O2
Formula Weight: 74.08
Flash Point: 51°(123°F)
Refractive Index: 1.3875
Odor: Unpleasant
Melting Point: -21°
Boiling Point: 140-141°
Beilstein Reference: 506071
Merck Reference: 14,7825
UN No.: 3463
MDL No.: MFCD00002756
Formula:
C₃H₆O₂ Boiling Pt: 140…141 °C Melting Pt: –21 °C Density: 0.992 Flash Pt: 51 °C (123 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002756 CAS Number: 79-09-4 EINECS: 201-176-3 UN: 3463 ADR: 8,II REACH: 01-2119486971-24 Merck Index: 14,7825 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...