2,3-Dimethyl-1,3-butadiene 98% stabilized

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,3-Dimethylbuta-1,3-diene

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 100 ppm

L04207-09 L04207-18
CAAAAL04207-09EA 123.71 CAD
CAAAAL04207-09 CAAAAL04207-18
2,3-Dimethyl-1,3-butadiene 98% stabilized
2,3-Dimethyl-1,3-butadiene

Purity: 98% CAS No.: 513-81-5 Molecular Formula: C6H10 Formula Weight: 82.15 Storage Temperature: 2°C to 8°C Flash Point: -1°(30°F) Refractive Index: 1.4380 Melting Point: -76° Boiling Point: 68-69° Beilstein Reference: 605285 Merck Reference: 14,3238 UN No.: 3295 MDL No.: MFCD00008595

Formula: C₆H₁₀
MW: 82.15 g/mol
Boiling Pt: 68…69 °C
Melting Pt: –76 °C
Density: 0.726
Flash Pt: –1 °C (30 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008595
CAS Number: 513-81-5
EINECS: 208-172-0
UN: 3295
ADR: 3,III
Merck Index: 14,3238

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR