1-Chloro-2-methylpropane 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L04181-18
L04181-30
CAAAAL04181-18EA
91.32
CAD
CAAAAL04181-18
CAAAAL04181-30
1-Chloro-2-methylpropane 98%
1-Chloro-2-methylpropane
Purity: 98%
CAS No.: 513-36-0
Molecular Formula: C4H9Cl
Formula Weight: 92.57
Flash Point: -7°(19°F)
Refractive Index: 1.3975
Odor: Sweet
Melting Point: -131°
Boiling Point: 68-69°
Beilstein Reference: 635650
Merck Reference: 14,5138
UN No.: 1127
MDL No.: MFCD00000954
Formula:
C₄H₉Cl Boiling Pt: 68…69 °C Melting Pt: –131 °C Density: 0.879 Flash Pt: –7 °C (19 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000954 CAS Number: 513-36-0 EINECS: 208-157-9 UN: 1127 ADR: 3,II Merck Index: 14,5138 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...