Citraconic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Methyl-2-butenedioic acid

L04178-09 L04178-18
CAAAAL04178-09EA 55.46 CAD
CAAAAL04178-09 CAAAAL04178-18
Citraconic acid 98+%
Citraconic acid

Purity: 98+% CAS No.: 498-23-7 Molecular Formula: C5H6O4 Formula Weight: 130.10 Melting Point: 88-91° Beilstein Reference: 1722679 Merck Reference: 14,2321 MDL No.: MFCD00078086

Formula: C₅H₆O₄
MW: 130.1 g/mol
Boiling Pt: 336 °C (1013 hPa)
Melting Pt: 88…91 °C
Density: 1.620
Storage Temperature: Ambient
MDL Number: MFCD00078086
CAS Number: 498-23-7
EINECS: 207-858-7
Merck Index: 14,2321

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR