1,1,1,2,2,3,3-Heptafluoro-4-iodobutane 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1H,1H-Heptafluorobutyl iodide, 2,2,3,3,4,4,4-Heptafluoro-1-iodobutane

L04173-03 L04173-06
CAAAAL04173-03EA 73 CAD
CAAAAL04173-03 CAAAAL04173-06
1,1,1,2,2,3,3-Heptafluoro-4-iodobutane 96%
1,1,1,2,2,3,3-Heptafluoro-4-iodobutane

Purity: 96% CAS No.: 374-98-1 Molecular Formula: C4H2F7I Formula Weight: 309.95 Flash Point: None Refractive Index: 1.3625 Boiling Point: 91-92° Beilstein Reference: 1761927 MDL No.: MFCD00013719

Formula: C₄H₂F₇I
MW: 309.95 g/mol
Boiling Pt: 91…92 °C
Density: 2.054
Storage Temperature: Ambient
MDL Number: MFCD00013719
CAS Number: 374-98-1
EINECS: 206-779-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR