4-Chloro-o-cresol 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Chloro-4-hydroxy-3-methylbenzene, 4-Chloro-2-methylphenol

L04164-06 L04164-14
CAAAAL04164-06EA 86.27 CAD
CAAAAL04164-06 CAAAAL04164-14
4-Chloro-o-cresol 97%
4-Chloro-o-cresol

Purity: 97% CAS No.: 1570-64-5 Molecular Formula: C7H7ClO Formula Weight: 142.59 Flash Point: 78°(172°F) Melting Point: 45-48° Boiling Point: 220-225° Beilstein Reference: 1906684 UN No.: 3437 MDL No.: MFCD00002321

Formula: C₇H₇ClO
MW: 142.58 g/mol
Boiling Pt: 224
Melting Pt: 45…48 °C
Density: 1.20
Flash Pt: 78 °C (172 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002321
CAS Number: 1570-64-5
EINECS: 216-381-3
UN: 3437
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR