2',4',6'-Trihydroxyacetophenone, hydrate 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L04156-06
L04156-14
CAAAAL04156-06EA
68.89
CAD
CAAAAL04156-06
CAAAAL04156-14
2',4',6'-Trihydroxyacetophenone, hydrate 98+%
2',4',6'-Trihydroxyacetophenone
Purity: 98+%
CAS No.: 480-66-0
Molecular Formula: C8H8O4.xH2O
Formula Weight: 168.15(anhy)
Melting Point: 218-223°
Beilstein Reference: 1911197
Formula:
C₈H₈O₄ Boiling Pt: 394 °C (1013 hPa) Melting Pt: 218…223 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149091 CAS Number: 480-66-0 EINECS: 207-556-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...