1-Octadecanethiol 96%, technical

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: n-Octadecyl mercaptan, n-Octadecanethiol, Stearyl mercaptane, 1-Mercaptooctadecane, Octadecane-1-thiol

L04123-14 L04123-22
CAAAAL04123-14EA 46.77 CAD
CAAAAL04123-14 CAAAAL04123-22
1-Octadecanethiol 96%, technical
1-Octadecanethiol

Purity: 96% CAS No.: 2885-00-9 Molecular Formula: CH3(CH2)17SH Formula Weight: 286.57 Flash Point: 185°(365°F) Odor: Stench Boiling Point: 180°/5mm Beilstein Reference: 1811934 MDL No.: MFCD00004886

Formula: C₁₈H₃₈S
MW: 286.57 g/mol
Boiling Pt: 204
Melting Pt: 30…33 °C
Density: 0.847
Flash Pt: 185 °C (365 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004886
CAS Number: 2885-00-9
EINECS: 220-744-1
UN: 3335
ADR: 9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR