N-Ethylisopropylamine 98%

Supplier: Thermo Scientific Chemicals
Danger

L04089-03 L04089-06
CAAAAL04089-03EA 51.19 CAD
CAAAAL04089-03 CAAAAL04089-06
N-Ethylisopropylamine 98%
N-Ethylisopropylamine

Purity: 98% CAS No.: 19961-27-4 Molecular Formula: C5H13N Formula Weight: 87.17 Flash Point: 16°(60°F) Refractive Index: 1.3920 Boiling Point: 71-73° Beilstein Reference: 505952 UN No.: 2733 MDL No.: MFCD00015105

Formula: C₅H₁₃N
Boiling Pt: 71…73 °C
Density: 0.720
Flash Pt: 16 °C (60 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015105
CAS Number: 19961-27-4
EINECS: 243-442-1
UN: 2733
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR