7-Hydroxycoumarin 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Umbelliferone, 7-Hydroxy-2H-chromen-2-one, 7-Hydroxy-2-benzopyrone
L04082-14
L04082-22
CAAAAL04082-14EA
113.92
CAD
CAAAAL04082-14
CAAAAL04082-22
7-Hydroxycoumarin 98%
7-Hydroxycoumarin
Purity: 98%
CAS No.: 93-35-6
Molecular Formula: C9H6O3
Formula Weight: 162.14
Melting Point: 228-232°
Beilstein Reference: 127683
Merck Reference: 14,9847
MDL No.: MFCD00006878
Formula:
C₉H₆O₃ MW: 162.14 g/mol Boiling Pt: 382 °C (1013 hPa) Melting Pt: 228…232 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006878 CAS Number: 93-35-6 EINECS: 202-240-3 Merck Index: 14,9847 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...