Methyltriethoxysilane 98%

Supplier: Thermo Scientific Chemicals
Warning

L04059-AK L04059-AU
CAAAAL04059-AKEA 72.84 CAD
CAAAAL04059-AK CAAAAL04059-AU
Methyltriethoxysilane 98%
Methyltriethoxysilane

Purity: 98% CAS No.: 2031-67-6 Molecular Formula: C7H18O3Si Formula Weight: 178.30 Flash Point: 23°(73°F) Refractive Index: 1.3835 Odor: Mild Melting Point: -50° Boiling Point: 141-143° Beilstein Reference: 1742453 UN No.: 1993 MDL No.: MFCD00009066

Formula: C₇H₁₈O₃Si
Boiling Pt: 141…143 °C
Melting Pt: < –50 °C
Density: 0.897
Flash Pt: 23 °C(73 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009066
CAS Number: 2031-67-6
EINECS: 217-983-9
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR