4-Chloro-1-butanol tech. 85%, Technical Grade

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Chlorobutan-1-ol

L04042-18 L04042-30
CAAAAL04042-18EA 120.71 CAD
CAAAAL04042-18 CAAAAL04042-30
4-Chloro-1-butanol tech. 85%, Technical Grade
4-Chloro-1-butanol

Purity: 85% CAS No.: 928-51-8 Molecular Formula: C4H9ClO Formula Weight: 108.57 Flash Point: 66°(151°F) Refractive Index: 1.4570 Boiling Point: 50-52°/1mm Beilstein Reference: 1731408 MDL No.: MFCD00002967

Formula: C₄H₉ClO
MW: 108.57 g/mol
Boiling Pt: 50…52 °C/1 mm
Density: 1.089
Flash Pt: 36 °C (97 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002967
CAS Number: 928-51-8
EINECS: 213-175-5
UN: 2924
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR