Isobutyric acid 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Methylpropionic acid, 2-Methylpropanoic acid
L04038-0F
L04038-AE
L04038-AP
CAAAAL04038-0FEA
167.48
CAD
CAAAAL04038-0F
CAAAAL04038-AE
CAAAAL04038-AP
Isobutyric acid 99%
Isobutyric acid
Purity: 99%
CAS No.: 79-31-2
Molecular Formula: (CH3)2CHCO2H
Formula Weight: 88.11
Flash Point: 59°(138°F)
Refractive Index: 1.3928
Odor: Pungent
Melting Point: -47°
Boiling Point: 153-154°
Beilstein Reference: 635770
Merck Reference: 14,5155
UN No.: 2529
MDL No.: MFCD00002658
Formula:
C₄H₈O₂ MW: 88.11 g/mol Boiling Pt: 155 Melting Pt: -45 Density: 0.948 Flash Pt: 59 °C (138 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002658 CAS Number: 79-31-2 EINECS: 201-195-7 UN: 2529 ADR: 3 III Merck Index: 14,5155 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...