Dimethoxy(diphenyl)silane 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L04031-0B
L04031-18
L04031-30
CAAAAL04031-0BEA
812.12
CAD
CAAAAL04031-0B
CAAAAL04031-18
CAAAAL04031-30
Dimethoxy(diphenyl)silane 97%
Dimethoxy(diphenyl)silane
Purity: 97%
CAS No.: 6843-66-9
Molecular Formula: C14H16O2Si
Formula Weight: 244.37
Flash Point: 121°(249°F)
Refractive Index: 1.5420
Boiling Point: 160-162°/15mm
Beilstein Reference: 2940458
MDL No.: MFCD00025690
Formula:
C₁₄H₁₆O₂Si Boiling Pt: 160…162 °C/15 mm Density: 1.079 Flash Pt: 121 °C(249 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00025690 CAS Number: 6843-66-9 EINECS: 229-929-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...