1,4-Butanesultone 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L03992-14
L03992-22
CAAAAL03992-14EA
82.32
CAD
CAAAAL03992-14
CAAAAL03992-22
1,4-Butanesultone 99%
1,4-Butanesultone
Purity: 99%
CAS No.: 1633-83-6
Molecular Formula: C4H8O3S
Formula Weight: 136.17
Flash Point: 153°(307°F)
Refractive Index: 1.4640
Melting Point: 12-15°
Boiling Point: 114-116°/1.5mm
Beilstein Reference: 110588
MDL No.: MFCD00006584
Formula:
C₄H₈O₃S Boiling Pt: 114…116 °C/1.5 mm Melting Pt: 12…15 °C Density: 1.335 Flash Pt: 153 °C(307 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006584 CAS Number: 1633-83-6 EINECS: 216-647-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...