Tetraethylammonium hydroxide 20% in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tetrylammonium hydroxide

L03968-22 L03968-36
CAAAAL03968-22EA 69.05 CAD
CAAAAL03968-22 CAAAAL03968-36
Tetraethylammonium hydroxide 20% in aqueous solution
Tetraethylammonium hydroxide

CAS No.: 77-98-5 Molecular Formula: C8H21NO Formula Weight: 147.26 Refractive Index: 1.3730 Beilstein Reference: 3912961 Merck Reference: 14,9201 MDL No.: MFCD00009024

Formula: C₈H₂₁NO
MW: 147.26 g/mol
Density: 1.023
Storage Temperature: Ambient
MDL Number: MFCD00009024
CAS Number: 77-98-5
EINECS: 201-073-3
UN: 2922
ADR: 8,III
Merck Index: 14,9201

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR