2,3-Dimercapto-1-propanol 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Dimercapto-2,3 propanol-1, Dithioglycerine, 2,3-Dimercaptopropanol, BAL, Dimercaprol, 2,3-Dimercaptopropan-1-ol, 2,3-Dimercaptopropane-1-ol

L03953-03 L03953-06
CAAAAL03953-03EA 112.81 CAD
CAAAAL03953-03 CAAAAL03953-06
2,3-Dimercapto-1-propanol 97%
2,3-Dimercapto-1-propanol

British Anti-Lewisite. A chelator that inhibits sugar nucleotide degradation.


Purity: 97% CAS No.: 59-52-9 Molecular Formula: C3H8OS2 Formula Weight: 124.22 Flash Point: 110°(230°F) Physical Form: Liquid Density: 1.244 Refractive Index: 1.5740 Sensitivity: Air Sensitive Appearance: Colorless to yellow Odor: Garlic-like Boiling Point: 120°/15mm Beilstein Reference: 1732058 Merck Reference: 14,3209 UN No.: 2810 MDL No.: MFCD00004864


British Anti-Lewisite. A chelator that inhibits sugar nucleotide degradation.

Formula: C₃H₈OS₂
MW: 124.22 g/mol
Boiling Pt: 120 °C/15 mm
Melting Pt: 77 °C
Density: 1.244
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004864
CAS Number: 59-52-9
EINECS: 200-433-7
UN: 2810
ADR: 6.1,III
Merck Index: 14,3209
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=745c14e3244d75cab02b536ae9d18ec21829aeba&vid=345ebd39845f4c372f03844bee87b4d8c249ec2b&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR