L(+)-Norleucine 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: H-Nle-OH, H-NorLeu-OH, (S)-2-Aminohexanoic acid, (S)-(+)-Norleucine

L03913-03 L03913-06
CAAAAL03913-03EA 67.94 CAD
CAAAAL03913-03 CAAAAL03913-06 CAAAAL03913-14
L(+)-Norleucine 99%
L(+)-Norleucine

Purity: 99% CAS No.: 327-57-1 Molecular Formula: C6H13NO2 Formula Weight: 131.18 Melting Point: 300° Beilstein Reference: 1721750 Merck Reference: 14,6706 Optical Rotation: +23° (c=5 in 5N HCl) MDL No.: MFCD00064423

Formula: C₆H₁₃NO₂
Boiling Pt: 246 °C (1013 hPa)
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00064423
CAS Number: 327-57-1
EINECS: 206-321-4
Merck Index: 14,6706

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR