N-Methylformamide 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Formmethylamide, Formic acid methylamide

L03908-0B L03908-18 L03908-30
CAAAAL03908-0BEA 169.06 CAD
CAAAAL03908-0B CAAAAL03908-18 CAAAAL03908-30
N-Methylformamide 99%
N-Methylformamide

Purity: 99% CAS No.: 123-39-7 Molecular Formula: C2H5NO Formula Weight: 59.07 Flash Point: 98°(208°F) Refractive Index: 1.4320 Melting Point: -4° Boiling Point: 181-185° Beilstein Reference: 1098352 Merck Reference: 14,6076 MDL No.: MFCD00003280

Formula: C₂H₅NO
MW: 59.07 g/mol
Boiling Pt: 181…185 °C
Melting Pt: –4 °C
Density: 1.018
Flash Pt: 98 °C(208 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003280
CAS Number: 123-39-7
EINECS: 204-624-6
Merck Index: 14,6076

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR