(2-Hydroxyphenoxy)acetic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L03897-06
L03897-14
CAAAAL03897-06EA
101.44
CAD
CAAAAL03897-06
CAAAAL03897-14
(2-Hydroxyphenoxy)acetic acid 98+%
(2-Hydroxyphenoxy)acetic acid
Purity: 98+%
CAS No.: 6324-11-4
Molecular Formula: C8H8O4
Formula Weight: 168.15
Melting Point: 125-130°
Beilstein Reference: 1948566
Formula:
(C₆H₄OH)OCH₂COOH Melting Pt: 125…130 °C Storage Temperature: Ambient |
MDL Number:
MFCD00015454 CAS Number: 6324-11-4 EINECS: 228-684-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...