3-Methylindole 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L03890-06
L03890-14
CAAAAL03890-06EA
49.3
CAD
CAAAAL03890-06
CAAAAL03890-14
3-Methylindole 99%
3-Methylindole
Purity: 99%
CAS No.: 83-34-1
Molecular Formula: C9H9N
Formula Weight: 131.18
Flash Point: 132°(269°F)
Odor: Stench
Boiling Point: 265-268°
Beilstein Reference: 111296
Merck Reference: 14,8560
MDL No.: MFCD00005627
Formula:
C₉H₉N Boiling Pt: 265…268 °C Melting Pt: 94…97 °C Density: 1.003 g/cm³ (99.6 °C) Flash Pt: 132 °C (269 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005627 CAS Number: 83-34-1 EINECS: 201-471-7 UN: 3077 ADR: 9,III Merck Index: 14,8560 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...