(R)-(-)-sec-Butylamine 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L03889-03
L03889-06
L03889-MD
CAAAAL03889-03EA
358.66
CAD
CAAAAL03889-03
CAAAAL03889-06
CAAAAL03889-MD
(R)-(-)-sec-Butylamine 99%
(R)-(-)-sec-Butylamine
Purity: 99%
CAS No.: 13250-12-9
Molecular Formula: C4H11N
Formula Weight: 73.14
Flash Point: -19°(-2°F)
Refractive Index: 1.3930
Odor: Amine-like
pH: 12.3
Boiling Point: 63°
Beilstein Reference: 1718760
Merck Reference: 14,1544
Optical Rotation: -7.5° (neat)
UN No.: 2733
MDL No.: MFCD00064416
Formula:
C₄H₁₁N Boiling Pt: 63 °C Melting Pt: –104 °C Density: 0.722 Flash Pt: –19 °C (–2 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00064416 CAS Number: 13250-12-9 EINECS: 236-232-6 UN: 3286 ADR: 3,II Merck Index: 14,1544 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...