Hexadecan-16-olide 97%

Supplier: Thermo Scientific Chemicals

L03864-09 L03864-18
CAAAAL03864-09EA 121.03 CAD
CAAAAL03864-09 CAAAAL03864-18
Hexadecan-16-olide 97%
Hexadecan-16-olide

Purity: 97% CAS No.: 109-29-5 Molecular Formula: C16H30O2 Formula Weight: 254.42 Flash Point: 100°(212°F) Melting Point: 33-34° Beilstein Reference: 148631 MDL No.: MFCD00039668

Formula: C₁₆H₃₀O₂
Melting Pt: 33…34 °C
Flash Pt: >100 °C(212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039668
CAS Number: 109-29-5
EINECS: 203-662-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR