2-Octene cis- and trans-mixture 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L03862-06
L03862-14
CAAAAL03862-06EA
52.3
CAD
CAAAAL03862-06
CAAAAL03862-14
2-Octene cis- and trans-mixture 98%
2-Octene
Purity: 98%
CAS No.: 111-67-1
Molecular Formula: C8H16
Formula Weight: 112.22
Flash Point: 14°(57°F)
Refractive Index: 1.4150
Boiling Point: 123-125°
Beilstein Reference: 1719495
UN No.: 3295
MDL No.: MFCD00009532
Formula:
C₈H₁₆ MW: 112.22 g/mol Boiling Pt: 123…125 °C Melting Pt: –106.8 °C Density: 0.715 Flash Pt: 14 °C (57 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009532 CAS Number: 111-67-1 EINECS: 203-894-2 UN: 3295 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...