2-Ethylhexyl acrylate 98% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
L03854-0F
L03854-AE
L03854-AP
CAAAAL03854-0FEA
202.24
CAD
CAAAAL03854-0F
CAAAAL03854-AE
CAAAAL03854-AP
2-Ethylhexyl acrylate 98% stabilized
2-Ethylhexyl acrylate
Purity: 98%
CAS No.: 103-11-7
Molecular Formula: C11H20O2
Formula Weight: 184.28
Flash Point: 79°(174°F)
Refractive Index: 1.4360
Melting Point: -90°
Boiling Point: 130°/50mm
Beilstein Reference: 1765828
MDL No.: MFCD00009495
Formula:
C₁₁H₂₀O₂ Boiling Pt: 130 °C/50 mm Melting Pt: –90 °C Density: 0.885 Flash Pt: 79 °C(174 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009495 CAS Number: 103-11-7 EINECS: 203-080-7 UN: 3334 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...