Methyl-4-chlorobutyrate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L03734-14
L03734-22
CAAAAL03734-14EA
50.4
CAD
CAAAAL03734-14
CAAAAL03734-22
Methyl-4-chlorobutyrate 98%
Methyl 4-chlorobutyrate
Purity: 98%
CAS No.: 3153-37-5
Molecular Formula: Cl(CH2)3CO2CH3
Formula Weight: 136.58
Flash Point: 59°(138°F)
Refractive Index: 1.4320
Boiling Point: 175-176°
Beilstein Reference: 1745117
UN No.: 3272
MDL No.: MFCD00001003
Formula:
C₅H₉ClO₂ Boiling Pt: 175…176 °C Density: 1.125 Flash Pt: 59 °C(138 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001003 CAS Number: 3153-37-5 EINECS: 221-592-9 UN: 3272 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...