2,3-Lutidine 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,3-Dimethylpyridine
L03697-14
L03697-22
L03697-36
CAAAAL03697-14EA
51.19
CAD
CAAAAL03697-14
CAAAAL03697-22
CAAAAL03697-36
2,3-Lutidine 99%
2,3-Lutidine
Purity: 99%
CAS No.: 583-61-9
Molecular Formula: C7H9N
Formula Weight: 107.16
Flash Point: 50°(122°F)
Refractive Index: 1.5080
Odor: Unpleasant
Melting Point: -17°
Boiling Point: 160-161°
Beilstein Reference: 106418
UN No.: 2929
MDL No.: MFCD00009605
Formula:
C₇H₉N MW: 107.16 g/mol Boiling Pt: 160…161 °C Melting Pt: –17 °C Density: 0.948 Flash Pt: 50 °C (122 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009605 CAS Number: 583-61-9 EINECS: 209-514-1 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...