2-Methyl-1-phenyl-2-propanol 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L03679-22
L03679-36
CAAAAL03679-22EA
58.93
CAD
CAAAAL03679-22
CAAAAL03679-36
2-Methyl-1-phenyl-2-propanol 98+%
2-Methyl-1-phenyl-2-propanol
Purity: 98+%
CAS No.: 100-86-7
Molecular Formula: C10H14O
Formula Weight: 150.22
Flash Point: 82°(179°F)
Refractive Index: 1.5150
Melting Point: 23-25°
Boiling Point: 215°
Beilstein Reference: 1855608
MDL No.: MFCD00004465
Formula:
C₆H₅CH₂C(CH₃)(OH)CH₃ Boiling Pt: 215 °C Melting Pt: 23…25 °C Density: 0.974 Flash Pt: 82 °C(179 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004465 CAS Number: 100-86-7 EINECS: 202-896-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...