Ethylammonium chloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Aminoethane hydrochloride, Ethylamine hydrochloride

L03624-0B L03624-30
CAAAL03624-0BEA 507.18 CAD
CAAAL03624-0B CAAAL03624-30
Ethylammonium chloride 98%
Ethylammonium chloride

Caution: Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Formula: C₂H₈ClN
Melting Pt: 95…100 °C
Density: 1.220
Storage Temperature: Ambient
MDL Number: MFCD00012885
CAS Number: 557-66-4
EINECS: 209-182-8
Merck Index: 14,3762

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR