6-Chloro-o-cresol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L03617-06
L03617-14
CAAAAL03617-06EA
97.01
CAD
CAAAAL03617-06
CAAAAL03617-14
6-Chloro-o-cresol 98%
6-Chloro-o-cresol
Purity: 98%
CAS No.: 87-64-9
Molecular Formula: C7H7ClO
Formula Weight: 142.59
Flash Point: 72°(161°F)
Refractive Index: 1.5450
Boiling Point: 75-76°/12mm
Beilstein Reference: 774623
UN No.: 2669
MDL No.: MFCD00020000
Formula:
C₇H₇ClO Boiling Pt: 75…76 °C/12 mm Melting Pt: 0 °C Density: 1.201 Flash Pt: 72 °C (161 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00020000 CAS Number: 87-64-9 EINECS: 201-760-8 UN: 2669 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...