(±)-2-Amino-1-propanol 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L03608-06
L03608-14
CAAAAL03608-06EA
92.75
CAD
CAAAAL03608-06
CAAAAL03608-14
(±)-2-Amino-1-propanol 98%
(±)-2-Amino-1-propanol
Purity: 98%
CAS No.: 6168-72-5
Molecular Formula: C3H9NO
Formula Weight: 75.11
Flash Point: 83°(181°F)
Refractive Index: 1.4495
Melting Point: 8°
Boiling Point: 174-176°
Beilstein Reference: 1209234
UN No.: 2735
MDL No.: MFCD00008085
Formula:
C₃H₉NO Boiling Pt: 174…176 °C Melting Pt: 8 °C Density: 0.965 Flash Pt: 83 °C (181 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008085 CAS Number: 6168-72-5 EINECS: 228-207-3 UN: 2735 ADR: 8,III Merck Index: 12,00488 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...