Neopentyl bromide 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2,2-Dimethyl-1-propyl bromide, 1-Bromo-2,2-dimethylpropane
L03579-06
L03579-14
CAAAAL03579-06EA
109.02
CAD
CAAAAL03579-06
CAAAAL03579-14
Neopentyl bromide 98%
Neopentyl bromide
Purity: 98%
CAS No.: 630-17-1
Molecular Formula: (CH3)3CCH2Br
Formula Weight: 151.05
Flash Point: 6°(42°F)
Refractive Index: 1.4360
Boiling Point: 104-106°
Beilstein Reference: 1730989
UN No.: 1993
MDL No.: MFCD00000209
Formula:
C₅H₁₁Br MW: 151.05 g/mol Boiling Pt: 104…106 °C Density: 1.195 Flash Pt: 6 °C (42 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000209 CAS Number: 630-17-1 EINECS: 211-132-5 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...