Cyclopentanethiol 97%

Supplier: Thermo Scientific Chemicals
Warning

L03577-06 L03577-14
CAAAAL03577-06EA 60.67 CAD
CAAAAL03577-06 CAAAAL03577-14
Cyclopentanethiol 97%
Cyclopentanethiol

Purity: 97% CAS No.: 1679-07-8 Molecular Formula: C5H10S Formula Weight: 102.20 Flash Point: 25°(77°F) Refractive Index: 1.4900 Odor: Stench Boiling Point: 131-132° Beilstein Reference: 2343861 UN No.: 3336 MDL No.: MFCD00001369

Formula: C₅H₁₀S
Boiling Pt: 131…132 °C
Density: 0.957
Flash Pt: 25 °C (77 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001369
CAS Number: 1679-07-8
EINECS: 216-841-3
UN: 3336
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR