Hexyltrimethoxysilane 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L03576-06
L03576-14
CAAAAL03576-06EA
35.39
CAD
CAAAAL03576-06
CAAAAL03576-14
Hexyltrimethoxysilane 97%
Hexyltrimethoxysilane
Purity: 97%
CAS No.: 3069-19-0
Molecular Formula: CH3(CH2)5Si(OCH3)3
Formula Weight: 206.36
Flash Point: 62°(143°F)
Refractive Index: 1.4070
Boiling Point: 202-203°
Beilstein Reference: 2347551
MDL No.: MFCD00039801
Formula:
C₉H₂₂O₃Si Boiling Pt: 202…203 °C Density: 0,920 Flash Pt: 62 °C(143 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00039801 CAS Number: 3069-19-0 EINECS: 221-331-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...